Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042621.1 | NP_001036086.1 | 5 - 6 | 769 | 86 | |
XM_008761315.1 | XP_008759537.1 | 5 - 6 | 898 | 86 | ||
XM_008761316.1 | XP_008759538.1 | 5 - 6 | 895 | 86 | ||
GenBank mRNA | AY724476.1 | - | 5 - 6 | 769 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |