Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005761.1 | NP_001005761.1 | 8 - 9 | 1577 | 81 | |
GenBank mRNA | AF526269.1 | - | 8 - 9 | 1186 | 81 | |
BC083644.1 | - | 8 - 9 | 1577 | 7134963 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |