Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108842.1 | NP_001102312.1 | 1 - 2 | 229 | 94 | |
XM_006248191.2 | XP_006248253.1 | 1 - 2 | 504 | 94 | ||
XM_006248193.2 | XP_006248255.1 | 1 - 2 | 503 | 94 | ||
GenBank mRNA | BC162029.1 | - | 1 - 2 | 350 | 9100375 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |