Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100831.1 | NP_001094301.1 | 3 - 4 | 575 | 64 | |
GenBank mRNA | BC105894.1 | - | 3 - 4 | 340 | 64 | |
CD371338.1 | - | 3 - 4 | 585 | 64 | ||
FQ213223.1 | - | 3 - 4 | 375 | 64 | ||
FQ214314.1 | - | 3 - 4 | 372 | 64 | ||
FQ223219.1 | - | 3 - 4 | 418 | 64 | ||
FQ233693.1 | - | 3 - 4 | 406 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |