Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109121.2 | NP_001102591.1 | 9 - 10 | 1036 | 80 | |
GenBank mRNA | BC168933.1 | - | 9 - 10 | 1036 | 7106331 | 80 |
FQ221736.1 | - | 9 - 10 | 1112 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |