Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106352.1 | NP_001099822.1 | 6 - 7 | 1346 | 81 | |
XM_008760399.1 | XP_008758621.1 | 6 - 7 | 1181 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |