Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025010.1 | NP_001020181.1 | 3 - 4 | 224 | 62 | |
GenBank mRNA | BC089834.1 | - | 3 - 4 | 224 | 7325650 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |