Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213629.1 | NP_998794.1 | 3 - 4 | 432 | 102 | |
XM_006243047.2 | XP_006243109.1 | 3 - 4 | 443 | 102 | ||
GenBank mRNA | AY501475.1 | - | 3 - 4 | 432 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |