Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012747.2 | NP_001012765.1 | 1 - 2 | 104 | 75 | |
GenBank mRNA | AY917132.1 | - | 1 - 2 | 82 | 75 | |
BC103643.1 | - | 1 - 2 | 104 | 7301307 | 75 | |
FQ227943.1 | - | 1 - 2 | 152 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |