Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012496.2 | NP_036628.2 | 7 - 8 | 853 | 66 | |
GenBank mRNA | BC081697.1 | - | 7 - 8 | 853 | 7109764 | 66 |
FQ209819.1 | - | 7 - 8 | 883 | 66 | ||
FQ210453.1 | - | 5 - 6 | 580 | 66 | ||
FQ210849.1 | - | 7 - 8 | 875 | 66 | ||
FQ211293.1 | - | 5 - 6 | 551 | 66 | ||
FQ219077.1 | - | 5 - 6 | 557 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |