Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013074.1 | NP_001013092.1 | 6 - 7 | 796 | 73 | |
XM_006256187.2 | XP_006256249.1 | 7 - 8 | 1831 | 73 | ||
XM_006256188.2 | XP_006256250.1 | 8 - 9 | 1893 | 73 | ||
XM_006256189.2 | XP_006256251.1 | 8 - 9 | 1885 | 73 | ||
XM_006256190.2 | XP_006256252.1 | 7 - 8 | 1829 | 73 | ||
XM_008772808.1 | XP_008771030.1 | 8 - 9 | 1708 | 73 | ||
XM_008772809.1 | XP_008771031.1 | 7 - 8 | 1823 | 73 | ||
XM_008772810.1 | XP_008771032.1 | 7 - 8 | 1711 | 73 | ||
GenBank mRNA | BC091397.1 | - | 6 - 7 | 796 | 7323315 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |