Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106031.1 | NP_001099501.1 | 1 - 2 | 76 | 58 | |
XM_006251857.2 | XP_006251919.1 | 1 - 2 | 106 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |