Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108027.1 | NP_001101497.1 | 8 - 9 | 819 | 101 | |
XM_006240169.2 | XP_006240231.1 | 8 - 9 | 724 | 101 | ||
XM_008764716.1 | XP_008762938.1 | 8 - 9 | 724 | 101 | ||
XM_008764717.1 | XP_008762939.1 | 8 - 9 | 632 | 101 | ||
GenBank mRNA | BC159403.1 | - | 8 - 9 | 730 | 7457550 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |