Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008386.1 | NP_001008387.1 | 4 - 5 | 875 | 79 | |
XM_008764603.1 | XP_008762825.1 | 4 - 5 | 2226 | 79 | ||
GenBank mRNA | AY769991.1 | - | 3 - 4 | 560 | 79 | |
AY769992.1 | - | 4 - 5 | 875 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |