Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014196.1 | NP_001014218.1 | 6 - 7 | 678 | 106 | |
XM_008764660.1 | XP_008762882.1 | 5 - 6 | 564 | 106 | ||
GenBank mRNA | BC079257.1 | - | 6 - 7 | 678 | 7133341 | 106 |
FQ227548.1 | - | 5 - 6 | 535 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |