Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761269.1 | XP_008759491.1 | 1330 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons |