Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107943.1 | NP_001101413.1 | 2 - 3 | 311 | 97 | |
XM_006238363.2 | XP_006238425.1 | 2 - 3 | 353 | 97 | ||
GenBank mRNA | FQ229191.1 | - | 2 - 3 | 286 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |