Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024366.1 | NP_001019537.1 | 4 - 5 | 540 | 71 | |
XM_008773430.1 | XP_008771652.1 | 4 - 5 | 1061 | 71 | ||
GenBank mRNA | BC079325.1 | - | 4 - 5 | 540 | 7135460 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |