Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012869.1 | NP_037001.1 | 2 | 1550 | 107 | |
XM_006253013.2 | XP_006253075.1 | 2 | 1405 | 107 | ||
XM_006253014.2 | XP_006253076.1 | 3 | 1595 | 107 | ||
GenBank mRNA | U56078.1 | - | 1 | 1367 | 107 | |
U66274.1 | - | 2 | 1550 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |