Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191550.1 | NP_001178479.1 | 7 | 940 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Amplicon spans exons and probe does not span exons |