Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250250.2 | XP_006250312.1 | 7 | 5216 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |