Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009271.1 | NP_001009271.1 | 7 - 8 | 880 | 178 | |
XM_008770986.1 | XP_008769208.1 | 7 - 8 | 2922 | 178 | ||
GenBank mRNA | AY569011.1 | - | 7 - 8 | 873 | 178 | |
BC088431.1 | - | 7 - 8 | 880 | 7124640 | 178 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |