Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009486.1 | NP_001009486.1 | 2 - 3 | 263 | 116 | |
GenBank mRNA | AY649835.1 | - | 2 - 3 | 263 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |