Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009710.1 | NP_001009710.1 | 6 - 7 | 1941 | 156 | |
XM_006238682.2 | XP_006238744.1 | 6 - 7 | 2326 | 156 | ||
GenBank mRNA | BC087037.1 | - | 6 - 7 | 1941 | 7133544 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |