Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008811.1 | NP_001008811.1 | 1 - 2 | 426 | 140 | |
XM_006242351.2 | XP_006242413.1 | 1 - 2 | 482 | 140 | ||
GenBank mRNA | AB355640.1 | - | 1 - 2 | 426 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |