Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slpil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008872.1 | NP_001008872.1 | 1 - 2 | 85 | 136 | |
XM_008762532.1 | XP_008760754.1 | 1 - 2 | 85 | 136 |
Gene Symbol: LOC103694893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773874.1 | XP_008772096.1 | 1 - 2 | 85 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |