Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004099.1 | NP_001004099.1 | 2 | 2255 | 109 | |
GenBank mRNA | BC078952.1 | - | 2 | 2255 | 7110122 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |