Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 5 - 6 | 347 | 108 | |
GenBank mRNA | CB544748.1 | - | 5 - 6 | 358 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |