Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108799.2 | NP_001102269.1 | 1 - 2 | 386 | 78 | |
GenBank mRNA | BC168738.1 | - | 1 - 2 | 386 | 6918011 | 78 |
FQ211150.1 | - | 1 - 2 | 155 | 78 | ||
FQ211594.1 | - | 1 - 2 | 182 | 78 | ||
FQ216958.1 | - | 1 - 2 | 154 | 78 | ||
FQ218320.1 | - | 1 - 2 | 154 | 78 | ||
FQ221159.1 | - | 1 - 2 | 154 | 78 | ||
FQ221342.1 | - | 1 - 2 | 156 | 78 | ||
FQ222681.1 | - | 1 - 2 | 154 | 78 | ||
FQ222690.1 | - | 1 - 2 | 155 | 78 | ||
FQ223187.1 | - | 1 - 2 | 154 | 78 | ||
FQ228255.1 | - | 1 - 2 | 155 | 78 | ||
FQ228375.1 | - | 1 - 2 | 156 | 78 | ||
FQ228595.1 | - | 1 - 2 | 155 | 78 | ||
FQ229059.1 | - | 1 - 2 | 156 | 78 | ||
FQ232283.1 | - | 1 - 2 | 154 | 78 | ||
FQ232532.1 | - | 1 - 2 | 157 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |