Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134702.1 | NP_001128174.1 | 9 - 10 | 1022 | 71 | |
XM_006251529.2 | XP_006251591.1 | 9 - 10 | 2270 | 71 | ||
GenBank mRNA | BC161984.1 | - | 9 - 10 | 1022 | 9030600 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |