Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212509.2 | NP_997674.1 | 3 - 4 | 658 | 97 | |
XM_006256046.2 | XP_006256108.1 | 3 - 4 | 593 | 97 | ||
GenBank mRNA | BC099772.1 | - | 3 - 4 | 602 | 7932537 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |