Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107580.1 | NP_001101050.1 | 1 - 2 | 22 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |