Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203333.2 | NP_976078.1 | 3 | 1042 | 123 | |
GenBank mRNA | BC062401.1 | - | 3 | 1003 | 5621455 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |