Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191949.1 | NP_001178878.1 | 13 - 14 | 1577 | 76 | |
XM_006229401.2 | XP_006229463.1 | 14 - 15 | 1768 | 76 | ||
XM_006229403.2 | XP_006229465.1 | 14 - 15 | 1713 | 76 | ||
XM_006229404.2 | XP_006229466.1 | 14 - 15 | 1768 | 76 | ||
XM_008759547.1 | XP_008757769.1 | 14 - 15 | 3568 | 76 | ||
XM_008759548.1 | XP_008757770.1 | 13 - 14 | 1644 | 76 | ||
XM_008759549.1 | XP_008757771.1 | 14 - 15 | 1717 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |