Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297557.1 | NP_001284486.1 | 4 - 5 | 631 | 74 | |
NM_001301048.1 | NP_001287977.1 | 4 - 5 | 625 | 74 | ||
NM_139081.2 | NP_620781.1 | 4 - 5 | 631 | 74 | ||
GenBank mRNA | D10706.1 | - | 4 - 5 | 603 | 74 | |
FQ213504.1 | - | 4 - 5 | 631 | 74 | ||
FQ213794.1 | - | 4 - 5 | 631 | 74 | ||
FQ214019.1 | - | 4 - 5 | 626 | 74 | ||
FQ214051.1 | - | 4 - 5 | 626 | 74 | ||
FQ216817.1 | - | 4 - 5 | 631 | 74 | ||
FQ218486.1 | - | 4 - 5 | 626 | 74 | ||
FQ218990.1 | - | 4 - 5 | 626 | 74 | ||
FQ220491.1 | - | 4 - 5 | 626 | 74 | ||
FQ220580.1 | - | 4 - 5 | 631 | 74 | ||
FQ225116.1 | - | 4 - 5 | 626 | 74 | ||
FQ225909.1 | - | 4 - 5 | 626 | 74 | ||
FQ225949.1 | - | 4 - 5 | 629 | 74 | ||
FQ230452.1 | - | 4 - 5 | 626 | 74 | ||
FQ231088.1 | - | 4 - 5 | 626 | 74 | ||
FQ232667.1 | - | 4 - 5 | 626 | 74 | ||
FQ232997.1 | - | 4 - 5 | 626 | 74 | ||
FQ233723.1 | - | 4 - 5 | 626 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |