Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001181176.3 | NP_011561.3 | 1 | 2857 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |