Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182692.1 | NP_013911.1 | 1 | 1252 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Inventoried Both primers and probe map within a single exon |