Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184113.1 | NP_015341.1 | 1 | 643 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |