Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927476.5 | XP_001927511.1 | 1096 | 122 | ||
GenBank mRNA | AB271955.1 | - | 5 - 6 | 948 | 122 | |
AK396352.1 | - | 5 - 6 | 1035 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |