Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925664.5 | XP_001925699.2 | 1653 | 98 | ||
XM_021075120.1 | XP_020930779.1 | 1659 | 98 | |||
GenBank mRNA | AF069646.1 | - | 1 - 2 | 80 | 98 | |
AK352011.1 | - | 13 - 14 | 1512 | 98 | ||
AK394001.1 | - | 13 - 14 | 1599 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |