Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113452.1 | NP_001106923.1 | 4 - 5 | 1739 | 75 | |
XM_005673012.3 | XP_005673069.1 | 1759 | 75 | |||
XM_005673013.3 | XP_005673070.1 | 1391 | 75 | |||
XM_013985461.2 | XP_013840915.1 | 2176 | 75 | |||
XM_013985462.2 | XP_013840916.1 | 993 | 75 | |||
XM_021077198.1 | XP_020932857.1 | 1683 | 75 | |||
XM_021077200.1 | XP_020932859.1 | 1391 | 75 | |||
GenBank mRNA | AF069995.1 | - | 2 - 3 | 281 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |