Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130211.1 | NP_001123683.1 | 4 - 5 | 748 | 146 | |
GenBank mRNA | AF311909.1 | - | 2 - 3 | 230 | 146 | |
AK231059.1 | - | 4 - 5 | 581 | 146 | ||
AK351736.1 | - | 4 - 5 | 561 | 146 | ||
AK391541.1 | - | 4 - 5 | 563 | 146 | ||
AY923074.1 | - | 4 - 5 | 748 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |