Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128456.1 | NP_001121928.1 | 2 - 3 | 198 | 110 | |
XM_005655480.3 | XP_005655537.1 | 528 | 110 | |||
XM_005655481.3 | XP_005655538.1 | 638 | 110 | |||
GenBank mRNA | AK240087.1 | - | 3 - 4 | 515 | 110 | |
AK398481.1 | - | 3 - 4 | 511 | 110 | ||
EU366153.1 | - | 2 - 3 | 198 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |