Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005660371.3 | XP_005660428.1 | 479 | 76 | ||
XM_005660372.3 | XP_005660429.1 | 581 | 76 | |||
XM_021067967.1 | XP_020923626.1 | 478 | 76 | |||
GenBank mRNA | AK233369.1 | - | 4 - 5 | 464 | 76 | |
AK346200.1 | - | 4 - 5 | 464 | 76 | ||
AK346379.1 | - | 4 - 5 | 464 | 76 | ||
M35990.1 | - | 4 - 5 | 370 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |