Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164007.1 | NP_001157479.1 | 2 - 3 | 357 | 86 | |
GenBank mRNA | AK231773.1 | - | 2 - 3 | 319 | 86 | |
AK233241.1 | - | 2 - 3 | 357 | 86 | ||
AK233706.1 | - | 2 - 3 | 330 | 86 | ||
AK392032.1 | - | 2 - 3 | 342 | 86 | ||
M86719.1 | - | 2 - 3 | 115 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |