Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244215.1 | NP_001231144.1 | 2 | 779 | 67 | |
GenBank mRNA | AF284498.1 | - | 1 | 552 | 67 | |
AK344310.1 | - | 2 | 779 | 67 | ||
BI341816.1 | - | 1 | 101 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |