Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003358932.4 | XP_003358980.1 | 6532 | 118 | ||
XM_003358933.4 | XP_003358981.1 | 6532 | 118 | |||
XM_005657147.3 | XP_005657204.1 | 6532 | 118 | |||
GenBank mRNA | AY072691.1 | - | 4 | 450 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |