Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244283.1 | NP_001231212.1 | 3 | 602 | 86 | |
XM_021093342.1 | XP_020949001.1 | 643 | 86 | |||
GenBank mRNA | AK391695.1 | - | 3 | 554 | 86 | |
AY349420.1 | - | 1 | 250 | 86 | ||
CF175298.1 | - | 1 | 434 | 86 | ||
EW464750.2 | - | 3 | 580 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |