Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY079212.1 | - | 1 | 248 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |