Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214417.1 | NP_999582.1 | 9 | 1450 | 75 | |
GenBank mRNA | AB052255.1 | - | 9 | 1461 | 75 | |
AK233106.1 | - | 9 | 1440 | 75 | ||
AK233296.1 | - | 9 | 1440 | 75 | ||
AK233494.1 | - | 9 | 1440 | 75 | ||
AY091516.1 | - | 9 | 1450 | 75 | ||
KP687255.1 | - | 9 | 1543 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |